Learn More
Thermo Scientific™ Nunc™ PETG Roller Bottles
Product Code. 10596895
Click to view available options
Cell Growth Area:
1050 cm2
1700 cm2
1800 cm2
2100 cm2
4200 cm2
Bottle Surface:
Pleated
Smooth
Packaging:
20/case
22/case
5/sleeve, 20/case
Unit Size:
Pack of 20
Pack of 22
Product Code. 10596895
Brand: Thermo Scientific™ 426022
Only null left
Add to basket
Add to basket
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor.
Increase cell expansion and product yield while eliminating the need to purchase additional production equipment or increase labor with Thermo Scientific™ Nunc™ PETG Roller Bottles. Virtually unbreakable PETG provides added assurance that valuable contents are safe during laboratory and industrial-scale applications including vaccines, cell culture expansion and production of other biologics.
Key Features:
  • Molded of durable PETG
  • Can be frozen down to -40°C — supports freeze thaw cell release methods, reducing reliance on trypsin
  • Provides solutions for all scales with wide range of surface areas from 1050cm2 to 4200cm2
  • Reduces wrist strain and increases productivity with quick-action ergonomic closure
  • Can be traced by lot number on every bottle
Recommended for:
  • Cell culture expansion
  • Vaccine production
  • Antibody production
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Pleated
Color Clear
Description 4260-22
Material PETG
No. per Case 22
No. per Pack 22
Packaging 22/case
Cell Growth Area 4200 cm2
Closure Included Yes
Graduated Yes
Shape Round
Product Type Roller Bottle
Closure Type Quick-Action Ergonomic
Show More Show Less
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.