missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ Nunc™ Edge™ 96-Well, Non-Treated, Flat-Bottom Microplate
58.40 GBP valid until 2024-06-30
Use promo code "21614" to get your promotional price.
Use promo code "21614" to get your promotional price.
Click to view available options
1/Pk., 50/Cs.
10/Pk., 160/Cs.
Description
Minimize plate evaporation and the risks associated with edge effect when using the uniquely engineered Thermo Scientific™ Nunc™ Edge 2.0 Plate in place of your standard 96-well plate. When filled with sterile water or media, the surrounding moat acts an evaporation buffer during extended incubation, yielding consistent, viable cells across the entire 96-well plate that in turn enhances productivity for cell-based assays. Enabling the use of all 96 wells allows researchers to plan experiments more efficiently, resulting in potentially less cost and related product waste.
Features
- The Edge 2.0 Plate provides an efficient and economical solution for preventing evaporation and in turn the risks associated with edge effect.
- The surrounding built-in moat is divided into 4 sectional reservoirs, each can be filled with 1.7 to 2.0mL sterile water, buffer or media.
- Replenish fluid in the reservoirs every 3 days in the humidified incubator to ensure the effectiveness of the evaporation barrier.
- Nunclon™ Delta cell culture surface treatment of the wells promotes cell attachment and growth.
- The non-treated surface is available for suspension cell culture or general solution-based assays
- Use the Edge 2.0 Plate in conjunction with the Thermo Scientific™ Varioskan™ LUX multimode microplate reader. The filled moat of the plate functions as passive humidity control, allowing for simultaneous incubation and signal detection during long-term applications with living cells inside the plate reader.
Specifications
Certifications/Compliance | All microplates manufactured by Nunc meet the recommendation of American National Standards (ANSI) (ANSI/SBS 1-2004) |
Lid | With Lid |
Culture Area | 0.35 cm2/well |
Array | 8 x 12 |
Packaging | 1/Pk., 50/Cs. |
Product Line | Nunc |
Color | Clear |
Product Type | Microplate |
Skirt Style | Low Flange |
Material | Polystyrene |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'