missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Nunc™ Cell Factory™ System Accessories

Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories.

Brand:  Thermo Scientific™ 173249

Code : 76

Additional Details : Weight : 0.17000kg

 View more versions of this product

Product Code. 10144302

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Further the development of applications that require a more closed cell culture system using Thermo Scientific™ Nunc™ Cell Factory™ Accessories. Reduce the need for open interventions and support the use of aseptic methods for fluid handing relating to filling, inoculation, feeding and harvesting of the Cell Factory system.

Connections Made Easy:
  • Plug a media bag or tubing set to any Cell Factory system for enhanced aseptic connectivity for filling and harvesting
Additional Features:
  • Save time and reduce costs
  • Minimize cleaning processes such as autoclaving and associated validations
  • Prevent build up of back pressure during filling application
Specifications

Specifications

Airvent Filter with HDPE Connector, 0.22µm
2
System Accessories
0.22μm
Cell Factory Systems (all models)
2
Sterile
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.