missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Macherey-Nagel™ NucleoSpin™ 8 Virus Strips
Additional 8-well strips that enable varying throughput when isolating viral RNA and DNA from serum, plasma, or cell-free biological fluids using the NucleoSpin™ 8 Virus full or core kit.
£58.80 - £205.00
Specifications
Format | 8-well Strip |
---|---|
Isolation Technology | Silica Membrane Technology |
Sample Type | Cell-free Biological Fluids, Plasma, Serum |
Efficiency | Binding capacity: 40 μg |
Elution Volume | Elution volume: 70 - 100 μL |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15840938
|
Macherey-Nagel™
740643.12S |
12 x 8-well Strips |
£58.80
Pack of 12 |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15850938
|
Macherey-Nagel™
740643.60S |
60 x 8-Well Strips |
£205.00
Pack of 60 |
Estimated Shipment: 09-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
NucleoSpin™ 8 Virus Kit—Isolate viral RNA and DNA from serum and plasma
- Parallel isolation using a flexible 8-well strip format for varying throughput
- For processing under vacuum or by centrifugation
- Suitable for manual and automated processing
- Innovative MN Wash Plate minimizes risk of cross-contamination
- Wide range of viral samples: HCV, HBV, HAV, HIV, HSV, HPV, VZV, EBV, parvovirus B19, H5N1, H1N1
- Typical downstream applications: PCR, RT-PCR, or any kind of enzymatic reaction
Specifications
8-well Strip | |
Cell-free Biological Fluids, Plasma, Serum | |
Elution volume: 70 - 100 μL | |
Parallel isolation of viral RNA and DNA | |
< 150 μL |
Silica Membrane Technology | |
Binding capacity: 40 μg | |
NucleoSpin 8 Virus full or core kit | |
8/Strip | |
60 min/6 strips |