missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel™ Nucleogel™ HPLC Column

Brand:  Macherey Nagel™ 719533

UNSPSC : 23151816

Code : 16

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 10496281

  • £1660.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications Description & Specifications

Specifications

Polymer-based
Anion exchange
Nucleotide Gel Anion I 7
HPLC Column
10μm
120mm
4 mM salicylate buffer pH 7.8
4.6mm
For Analysis
Polymer
Polymer-based
HPLC
1 to 14