missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nucleodur™ C18 Pyramid HPLC Columns

Brand:  Macherey-Nagel™ 760264.46

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12718732

  • £775.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

14%
Analytical
4.6mm
1mL/min.
100mm
Acetonitrile/water
Spherical
1 to 9
110Å
1 Each
340m2/g
L1
NUCLEODUR® C18 Pyramid
Octadecyl
Octadecyl
Yes
For Analytical and preparative separation of polar and midpolar compounds applications
Stainless Steel
Silica
3μm
NUCLEODUR® C18 Pyramid
0.9mL/g
Acetonitrile/water
EC HPLC Column
Analytical HPLC
Videos