missing translation for 'onlineSavingsMsg'
Learn More

Macherey-Nagel™ Nucleodur™ C18 Isis HPLC Column

Brand:  Macherey-Nagel™ 760397.46

Code : 16

Additional Details : Weight : 0.50000kg

 View more versions of this product

Product Code. 12747830

  • £675.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket

Specifications

Specifications

20%
Analytical
4.6mm
1mL/min.
75mm
Acetonitrile/water
Spherical
1 to 10
110Å
Acetonitrile/water
EC HPLC Column
Analytical HPLC
Polymeric octadecyl
Polymeric octadecyl
Yes
For Steroids, (o,p,m-) substituted aromatics, fat-soluble vitamins application
Stainless Steel
Silica
3μm
NUCLEODUR® C18 Isis
1 Each
340m2/g
L1
Nucleodur® C18 Isis
Videos