missing translation for 'onlineSavingsMsg'
Learn More

Macherey Nagel HPLC column 100mm x 4.6mm I.D. EC Isis particle size 5µm

Brand:  Macherey Nagel 760415.46

HPLC column 100mm x 4.6mm I.D. EC Isis particle size 5µm

Code : 16

Additional Details : Weight : 0.01000kg

Product Code. 12478457

  • £625.00 / Each


Call for latest availability
+44 (0)1509 555 500
Add to basket