Learn More
Nucleocapsid protein, Mouse anti-SARS-CoV-2, Clone: AS47, ACROBiosystems
HEK293 Monoclonal Antibody
Brand:  ACROBiosystems NUN-S47-10mg
Encompass_Preferred
Product Code. 17296380
  • £19860.00 / 10mg
Description
Description
This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is detected with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-L63, HCoV-OC43 and HCoV-HKU1.(Trehalose free)
Specifications
Specifications
Nucleocapsid protein
AS47
Trehalose free
10 mg
Primary
SARS-CoV-2
Recombinant Antibodies
Mouse IgG1 κ
ELISA
Unconjugated
HEK293
RUO
This product is a specific antibody against SARS-CoV-2 Nucleocapsid protein. No cross-reactivity is detected with nucleocapsid protein of other coronaviruses, including SARS-CoV, MERS-CoV, HCoV-229E, HCoV-L63, HCoV-OC43 and HCoV-HKU1.
2-8°C