missing translation for 'onlineSavingsMsg'
Learn More

NU 7026, Tocris Bioscience™

Selective DNA-PK inhibitor

Brand:  Tocris (Bio-Techne) 282810

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13441904

  • £162.36 / 10mg

Estimated Shipment
28-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

ATP-competitive inhibitor of DNA-dependent protein kinase (DNA-PK). Displays selectivity over other PIKK family enzymes (IC50 values are 0.23, 13.0, > 100 and > 100 μM for DNA-PK, PI3K, ATM and ATR respectively). Radiosensitizes both proliferating and quiescent mouse embryonic fibroblast cells to IR and inhibits DSB repair.
Specifications

Chemical Identifiers

154447-35-5
281.311
dna-pk inhibitor ii, 2-morpholin-4-yl-benzo h chromen-4-one, 2-morpholino-4h-benzo h chromen-4-one, 2-4-morpholinyl-4h-naphtho 1,2-b pyran-4-one, 2-4-morpholinyl-4h-naphthol 1,2-b pyran-4-one, 2-morpholin-4-ylbenzo h chromen-4-one, dna-dependent protein kinase inhibitor ii, 2-morpholin-4-yl benzo h chromen-4-one, d03tvf, d0w1st
2-morpholin-4-ylbenzo[h]chromen-4-one
C17H15NO3
KKTZALUTXUZPSN-UHFFFAOYSA-N
9860529
C1COCCN1C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3

Specifications

NU 7026
154447-35-5
dna-pk inhibitor ii, 2-morpholin-4-yl-benzo h chromen-4-one, 2-morpholino-4h-benzo h chromen-4-one, 2-4-morpholinyl-4h-naphtho 1,2-b pyran-4-one, 2-4-morpholinyl-4h-naphthol 1,2-b pyran-4-one, 2-morpholin-4-ylbenzo h chromen-4-one, dna-dependent protein kinase inhibitor ii, 2-morpholin-4-yl benzo h chromen-4-one, d03tvf, d0w1st
C1COCCN1C2=CC(=O)C3=C(O2)C4=CC=CC=C4C=C3
281.311
281.31
10mg
C17H15NO3
KKTZALUTXUZPSN-UHFFFAOYSA-N
2-morpholin-4-ylbenzo[h]chromen-4-one
9860529
>98%
Videos