missing translation for 'onlineSavingsMsg'
Learn More

NU 1025, Tocris Bioscience™

Potent PARP inhibitor

Brand:  Tocris (Bio-Techne) 140150

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439323

  • £547.32 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Novel, potent inhibitor of poly(ADP-ribose) polymerase (PARP). Ki and IC50 values are 48 and 400 nM respectively.
Specifications

Chemical Identifiers

90417-38-2
176.175
8-hydroxy-2-methylquinazoline-4-one, 8-hydroxy-2-methylquinazolin-4 3h-one, parp inhibitor vi, nu1025, 8-hydroxy-2-methyl-3h-quinazolin-4-one, 8-hydroxy-2-methyl-4 3h-quinazolinone, 8-hydroxy-2-methyl-3-hydro-quinazolin-4-one, 8-hydroxy-2-methylquinazolin-4 1h-one, 4pax, 8-hydroxy-2-methylquinazolin-4 3h-one, nu1
CHEBI:75399
CC1=NC(=O)C2=C(N1)C(=CC=C2)O
C9H8N2O2
YJDAOHJWLUNFLX-UHFFFAOYSA-N
63306
8-hydroxy-2-methyl-1H-quinazolin-4-one

Specifications

NU 1025
90417-38-2
8-hydroxy-2-methylquinazoline-4-one, 8-hydroxy-2-methylquinazolin-4 3h-one, parp inhibitor vi, nu1025, 8-hydroxy-2-methyl-3h-quinazolin-4-one, 8-hydroxy-2-methyl-4 3h-quinazolinone, 8-hydroxy-2-methyl-3-hydro-quinazolin-4-one, 8-hydroxy-2-methylquinazolin-4 1h-one, 4pax, 8-hydroxy-2-methylquinazolin-4 3h-one, nu1
CC1=NC(=O)C2=C(N1)C(=CC=C2)O
176.175
CHEBI:75399
>99%
50mg
C9H8N2O2
YJDAOHJWLUNFLX-UHFFFAOYSA-N
8-hydroxy-2-methyl-1H-quinazolin-4-one
63306
176.17
Videos