Learn More
Tocris Bioscience™ NTNCB hydrochloride
Potent and selective non-peptidic NPY Y5 antagonist
Brand:  Tocris Bioscience™ 215550
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13430684
  • £633.57 / 50mg
Estimated Shipment date
from Supplier 25-03-2020
Description
Description
Selective, non-peptide competitive NPY Y5 antagonist. Ki values are 8.0 and 16032 nM at human recombinant Y5 and Y1 receptors respectively. Potently antagonizes NPY inhibition of forskolin-stimulated cAMP. Displays some affinity for cloned human D2 and α2C receptors (Ki values are 63 and 100 nM respectively).
Specifications
Specifications
NTNCB hydrochloride
>99
50mg
NTNCB hydrochloride
508.07