missing translation for 'onlineSavingsMsg'
Learn More

Sino Biological™ NT5E Recombinant Human Protein, His Tag

Expressed from DNA sequence encoding human NT5E (Met 1-Lys 547) , w/o propeptide, fused to polyhistidine tag at C-terminus

Brand:  Sino Biological™ 10904H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15207375

  • £657.92 / 50µg


Estimated Shipment
17-05-2018
Add to basket

Description

Description

  • Measured by its ability to hydrolyze 5' phosphate group from substrate adenosine 5' monophosphate (AMP
  • Specific activity is >20,000 pmoles/min/μg
  • Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
Specifications

Specifications

Analysis of protein structure, used as immunogen, protein standard, or in cell biology research applications
4907
≥90% by SDS-PAGE
Human
<1 EU/μg
NT5E
CD Proteins
NT5E Recombinant Human Protein
Lyophilized
59.2kd
50μg
Full Length, Recombinant
5'-Nucleotidase, ECTO
Assay Standard
Mammalian
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.