missing translation for 'onlineSavingsMsg'
Learn More

NSC 632839 hydrochloride, Tocris Bioscience™

Inhibitor of ubiquitin isopeptidase activity

Brand:  Tocris (Bio-Techne) 264750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13461644

  • £374.89 / 50mg


Estimated Shipment
04-03-2019
Add to basket

Description

Description

Inhibitor of ubiquitin isopeptidase activity that displays no effect on the proteolytic activity of the proteasome. Induces apoptosis via a Bcl-2-dependent and apoptosome-independent pathway of caspase activation (IC50 values are 15.65 and 16.23 μM in E1A and E1A/C9CN cells respectively).
Specifications

Chemical Identifiers

157654-67-6
339.863
nsc 632839 hydrochloride, 3z,5z-3,5-bis 4-methylphenyl methylidene piperidin-4-one hydrochloride
(3Z,5E)-3,5-bis[(4-methylphenyl)methylidene]piperidin-4-one;hydrochloride
C21H22ClNO
ZOKZLTXPTLIWOJ-DKUXUYHUSA-N
21141595
CC1=CC=C(C=C1)C=C2CNCC(=CC3=CC=C(C=C3)C)C2=O.Cl

Specifications

NSC 632839 hydrochloride
157654-67-6
nsc 632839 hydrochloride, 3z,5z-3,5-bis 4-methylphenyl methylidene piperidin-4-one hydrochloride
CC1=CC=C(C=C1)C=C2CNCC(=CC3=CC=C(C=C3)C)C2=O.Cl
339.863
339.86
50mg
C21H22ClNO
ZOKZLTXPTLIWOJ-DKUXUYHUSA-N
(3Z,5E)-3,5-bis[(4-methylphenyl)methylidene]piperidin-4-one;hydrochloride
21141595
>99%
Videos