Learn More
Tocris Bioscience™ NSC 23766
Selective inhibitor of Rac1-GEF interaction; antioncogenic
Brand:  Tocris Bioscience™ 2161/10
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13460694
  • £157.29 / 10mg
Estimated Shipment date
from Supplier 03-04-2020
Description
Description
Selective inhibitor of Rac1-GEF interaction. Prevents Rac1 activation by Rac-specific guanine nucleotide exchange factors (GEFs) TrioN and Tiam1 (IC50 ∽ 50 μM) without affecting Cdc42 or RhoA activation. Inhibits Rac1-mediated cell functions and is reported to reverse tumor cell phenotypes in prostate cancer cells.
Specifications
Specifications
NSC 23766
>96
10mg
Rho GTPase Inhibitors
530.97