Learn More
Tocris Bioscience™ NSC 146109 hydrochloride
Cell-permeable, genotype-selective antitumor agent; activates p53-dependent transcription
Brand:  Tocris Bioscience™ 218510
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13430744
  • £161.70 / 10mg
Estimated Shipment date
from Supplier 31-03-2020
Description
Description
Cell-permeable, genotype-selective antitumor agent that activates p53-dependent transcription. Increases levels of endogenous p53 in tumor cells and protects p53 from Mdm2-mediated degradation. Displays some selectivity for tumor cells vs. normal cells in an MTT cell viability assay.
Specifications
Specifications
NSC 146109 hydrochloride
>99
10mg
NSC 146109 hydrochloride
316.85