Learn More
Tocris Bioscience™ NSC 146109 hydrochloride
Cell-permeable, genotype-selective antitumor agent; activates p53-dependent transcription
POA
Specifications
Purity >99%
View More Specs
Description
Description
Cell-permeable, genotype-selective antitumor agent that activates p53-dependent transcription. Increases levels of endogenous p53 in tumor cells and protects p53 from Mdm2-mediated degradation. Displays some selectivity for tumor cells vs. normal cells in an MTT cell viability assay.
Specifications
Specifications
>99%