Learn More
Tocris Bioscience™ NS 398
Cyclooxygenase-2 (COX-2) inhibitor
Brand:  Tocris Bioscience™ 094250
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13468183
  • £710.01 / 50mg
Estimated Shipment date
from Supplier 30-03-2020
Description
Description
Selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.
Specifications
Specifications
N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide
>99%
Observed MW: 314.36
Soluble to 100mM in DMSO
123653-11-2
C13H18N2O5S