missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ NS 398
Cyclooxygenase-2 (COX-2) inhibitor
POA
Specifications
Chemical Name or Material | N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide |
---|---|
CAS | 123653-11-2 |
Assay Percent Range | >99% |
Solubility | Soluble to 100mM in DMSO |
Molecular Formula | C13H18N2O5S |
Description
Selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100 μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.Specifications
N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide | |
>99% | |
C13H18N2O5S |
123653-11-2 | |
Soluble to 100mM in DMSO | |
Observed MW: 314.36 |
For Research Use Only