Learn More
Tocris Bioscience™ NS 398
Brand: Tocris Bioscience™ 0942/10
Code : Z1
Additional Details : Weight : 0.01000kg
Description
Selective cyclooxygenase-2 inhibitor (IC50 values are 3.8 and > 100μM for COX-2 and COX-1 respectively). Induces apoptosis in colorectal tumor cells and elevates COX-2 protein expression in vitro. Orally active and non-ulcerogenic analgesic and anti-inflammatory in vivo.Specifications
N-[2-Cyclohexyloxy-4-nitrophenyl]methanesulfonamide | |
>99% | |
Observed MW: 314.36 | |
Soluble to 100mM in DMSO |
123653-11-2 | |
C13H18N2O5S | |
10mg |
For Research Use Only