missing translation for 'onlineSavingsMsg'
Learn More

NS 309, Tocris Bioscience™

Positive modulator of KCa2 and KCa3.1 channels

Brand:  Tocris (Bio-Techne) 389510

Code : Z1

Additional Details : Weight : 0.01100kg

Quantity 10mg

 View more versions of this product

Product Code. 13463774

  • £95.85 / 10mg


Estimated Shipment
04-04-2019
Add to basket

Description

Description

Positive modulator of small- and intermediate- conductance Ca 2+ -activated K + channels (K Ca 2 and K Ca 3.1 channels); increases Ca 2+ sensitivity. Displays no activity at BK channels.
Specifications

Chemical Identifiers

18711-16-5
231.032
6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid
6,7-dichloro-3-(hydroxyamino)indol-2-one
C8H4Cl2N2O2
CVOUSAVHMDXCKG-UHFFFAOYSA-N
11637204
C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl

Specifications

NS 309
18711-16-5
6,7-dichloro-1h-indole-2,3-dione 3-oxime, 6,7-dichloro-3-hydroxyimino indolin-2-one, z-6,7-dichloro-3-hydroxyimino indolin-2-one, 6,7-dichloro-3-hydroxyamino indol-2-one, 3-oxime-6,7-dichloro-1h-indole-2,3-dione, 6,7-dichloro-3-hydroxyamino-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1,3-dihydro-2h-indol-2-one, 3e-6,7-dichloro-3-hydroxyimino-1h-indol-2-one, ns309 hplc, ns hplc , solid
C1=CC(=C(C2=NC(=O)C(=C21)NO)Cl)Cl
231.032
231.04
10mg
C8H4Cl2N2O2
CVOUSAVHMDXCKG-UHFFFAOYSA-N
6,7-dichloro-3-(hydroxyamino)indol-2-one
11637204
>98%
Videos