missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human NRL Antibody, R&D Systems™
Goat Polyclonal Antibody has been used in 2 publications
POA
Specifications
Antigen | NRL |
---|---|
Concentration | LYOPH |
Dilution | Western Blot 0.1 ug/mL |
Applications | Western Blot |
Classification | Polyclonal |
Description
NRL Polyclonal specifically detects NRL in Human samples. It is validated for Western Blot.Specifications
NRL | |
Western Blot 0.1 ug/mL | |
Polyclonal | |
Goat | |
Human | |
D14S46Eneural retina-specific leucine zipper protein, neural retina leucine zipper, NRL-MAF, RP27, RP27neural retinal-specific leucine zipper | |
NRL | |
IgG | |
Affinity Purified | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
LYOPH | |
Western Blot | |
Unconjugated | |
RUO | |
P54845 | |
4901 | |
E. coli-derived recombinant human NRL Met1-Leu237 Accession # P54845 | |
Primary | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Detects human NRL in direct ELISAs and Western blots. |
For Research Use Only