missing translation for 'onlineSavingsMsg'
Learn More

NPS 2390, Tocris Bioscience™

Group I mGlu antagonist

Brand:  Tocris (Bio-Techne) 413450

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13444214

  • £487.52 / 50mg

Estimated Shipment
09-07-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Group I mGlu antagonist; displays noncompetitive antagonist activity at both mGlu1 and mGlu5 receptors. Thought to act on a site separate from the glutamate binding pocket.
Specifications

Chemical Identifiers

226878-01-9
307.397
n-adamantan-1-yl quinoxaline-2-carboxamide, n-1-adamantyl quinoxaline-2-carboxamide, quinoxaline-2-carboxylic acid adamantan-1-ylamide, n-tricyclo 3.3.1.13,7 dec-1-yl-2-quinoxalinecarboxamide, d09cpy, d0w0xm, n-1-adamantyl-2-quinoxalinecarboxamide, nps hplc , solid, 2-quinoxalinecarboxamide, n-tricyclo 3.3.1.13,7 dec-1-yl, n-3r,5s,7s-adamantan-1-yl quinoxaline-2-carboxamide
N-(1-adamantyl)quinoxaline-2-carboxamide
C19H21N3O
ZKFVOZCCAXQXBU-UHFFFAOYSA-N
7067728
C1C2CC3CC1CC(C2)(C3)NC(=O)C4=NC5=CC=CC=C5N=C4

Specifications

NPS 2390
226878-01-9
n-adamantan-1-yl quinoxaline-2-carboxamide, n-1-adamantyl quinoxaline-2-carboxamide, quinoxaline-2-carboxylic acid adamantan-1-ylamide, n-tricyclo 3.3.1.13,7 dec-1-yl-2-quinoxalinecarboxamide, d09cpy, d0w0xm, n-1-adamantyl-2-quinoxalinecarboxamide, nps hplc , solid, 2-quinoxalinecarboxamide, n-tricyclo 3.3.1.13,7 dec-1-yl, n-3r,5s,7s-adamantan-1-yl quinoxaline-2-carboxamide
C1C2CC3CC1CC(C2)(C3)NC(=O)C4=NC5=CC=CC=C5N=C4
307.397
307.39
50mg
C19H21N3O
ZKFVOZCCAXQXBU-UHFFFAOYSA-N
N-(1-adamantyl)quinoxaline-2-carboxamide
7067728
>99%
Videos