missing translation for 'onlineSavingsMsg'
Learn More
Learn More
NPR-1 Goat anti-Mouse, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | NPRA/NPR1 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 Accession # P18293 |
Gene ID (Entrez) | 4881 |
Gene Accession No. | P18293 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
NPRA/NPR1 Polyclonal specifically detects NPRA/NPR1 in Mouse samples. It is validated for Western Blot.Specifications
NPRA/NPR1 | |
4881 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects mouse NPRA/NPR1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 50% cross-reactivity with recombinant human (rh) NPR1 is observed and 5% cross-reactivity with rhNPR2 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant mouse NPRA/NPR1 Ser29-Glu469 Accession # P18293 | |
P18293 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only