missing translation for 'onlineSavingsMsg'
Learn More

NPEC-caged-(S)-AMPA, Tocris Bioscience™

Caged (S)-AMPA

Brand:  Tocris (Bio-Techne) 384010

Code : Z1

Additional Details : Weight : 0.01000kg

Product Code. 13463684

  • £209.88 / 10mg


Estimated Shipment
29-03-2019
Add to basket

Description

Description

1-(2-nitrophenyl)ethyl caged version of (S)-AMPA (Cat. No. 0254), agonist at the AMPA receptor. Extinction coefficient (ε 347 ) = 660 M -1 cm -1 quantum yield (φ) = 0.64. Suitable for near-UV uncaging. No inhibition of GABA-A receptors at concentrations used experimentally.
Specifications

Specifications

NPEC-caged-(S)-AMPA
1257323-84-4
npec-caged-s-ampa, n-1-2-nitrophenyl ethylcarboxy-s-?-1-2-nitrophenyl ethylcarboxyamino-3-hydroxy-5-methyl-4-isoxazolepropionic acid
CC1=C(C(=O)NO1)CC(C(=O)O)NC(=O)OC(C)C2=CC=CC=C2[N+](=O)[O-]
379.325
379.32
10mg
C16H17N3O8
LGVJRWDYNJZRRP-MYIOLCAUSA-N
(2S)-3-(5-methyl-3-oxo-1,2-oxazol-4-yl)-2-[1-(2-nitrophenyl)ethoxycarbonylamino]propanoic acid
90488885
>98%
Videos