missing translation for 'onlineSavingsMsg'
Learn More

NPEC-caged-D-AP5, Tocris Bioscience™

Caged D-AP5 (Cat.No. 0106)

Brand:  RD Systems (Bio-Techne) 4230/10

Code : Z1

Additional Details : Weight : 0.00010kg

Quantity 10mg

 View more versions of this product

Product Code. 13464394

  • £184.80 / 10mg


Estimated Shipment
28-02-2019
Add to basket

Description

Description

D-AP5 (Cat.No. 0106) caged with the photosensitive 1-(2-nitrophenyl)ethoxycarbonyl (NPEC) group. NMDA receptor antagonist.
Specifications

Chemical Identifiers

1416943-27-5
390.285
npec-caged-d-ap5, d-1-2-nitrophenyl ethyl carbamoyl-2-amino-5-phosphonopentanoic acid
(2R)-2-[1-(2-nitrophenyl)ethoxycarbonylamino]-5-phosphonopentanoic acid
C14H19N2O9P
RLEANJAZNZWLHC-HCCKASOXSA-N
90488920
CC(C1=CC=CC=C1[N+](=O)[O-])OC(=O)NC(CCCP(=O)(O)O)C(=O)O

Specifications

NPEC-caged-D-AP5
1416943-27-5
npec-caged-d-ap5, d-1-2-nitrophenyl ethyl carbamoyl-2-amino-5-phosphonopentanoic acid
CC(C1=CC=CC=C1[N+](=O)[O-])OC(=O)NC(CCCP(=O)(O)O)C(=O)O
390.285
390.28
10mg
C14H19N2O9P
RLEANJAZNZWLHC-HCCKASOXSA-N
(2R)-2-[1-(2-nitrophenyl)ethoxycarbonylamino]-5-phosphonopentanoic acid
90488920
>99%
Videos