missing translation for 'onlineSavingsMsg'
Learn More

NPEC-caged-(1S,3R)-ACPD, Tocris Bioscience™

Caged (1S,3R)-ACPD

Brand:  Tocris (Bio-Techne) 333110

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13442824

  • £242.72 / 10mg

Estimated Shipment
14-09-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

(1S,3R)-ACPD (Cat.No. 0284) caged with the photosensitive 1-(2-nitrophenyl)ethoxycarbonyl (NPEC) group. Extinction coefficient (ε347) = 660 M-1cm-1, quantum yield (φ) = 0.64. Suitable for near-UV uncaging . No inhibition of GABA-A receptors at concentrations used experimentally.
Specifications

Chemical Identifiers

1315379-60-2
366.326
npec-caged-1s,3r-acpd, 1s,3r-1-2-nitrophenyl ethylcarboxyaminocyclopentane-1,3-dicarboxylic acid
(1S,3R)-1-[1-(2-nitrophenyl)ethoxycarbonylamino]cyclopentane-1,3-dicarboxylic acid
C16H18N2O8
SKBDCZAPZZBWJU-XOXYBHGNSA-N
90488832
CC(C1=CC=CC=C1[N+](=O)[O-])OC(=O)NC2(CCC(C2)C(=O)O)C(=O)O

Specifications

NPEC-caged-(1S, 3R)-ACPD
1315379-60-2
npec-caged-1s,3r-acpd, 1s,3r-1-2-nitrophenyl ethylcarboxyaminocyclopentane-1,3-dicarboxylic acid
CC(C1=CC=CC=C1[N+](=O)[O-])OC(=O)NC2(CCC(C2)C(=O)O)C(=O)O
366.326
366.32
10mg
C16H18N2O8
SKBDCZAPZZBWJU-XOXYBHGNSA-N
(1S,3R)-1-[1-(2-nitrophenyl)ethoxycarbonylamino]cyclopentane-1,3-dicarboxylic acid
90488832
>98%
Videos
Documents