Learn More
Invitrogen™ Novex™ Western-Star™ Immunodetection System
High sensitivity, high signal intensity, chemiluminescence-based immunodetection system
Brand:  Invitrogen™ T1046
Code : NEW
Additional Details : Weight : 0.00450kg
 View more versions of this product
Product Code. 12654295
  • £326.46 / Each
Estimated Shipment
15-03-2018
Add to basket
Specifications
Specifications
Mouse
Protein
30 blots
AP (Alkaline Phosphatase) Substrate
Western-SuperStar™ Reagent Set (containing Blocker, Wash Buffer, Assay Buffer, Substrate, Enhancer, Blot-Loc™ bags, Development Folders and pair of Tweezers) and anti-rabbit secondary antibody conjugated to alkaline phosphatase.