Learn More
Invitrogen™ Novex™ Western-Star™ Immunodetection System
High sensitivity, high signal intensity, chemiluminescence-based immunodetection system
Brand:  Invitrogen™ T1048
Code : NEW
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 12664295
  • £321.25 / Each
Estimated Shipment
06-02-2018
Add to basket
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Rabbit
Protein
30 blots
AP (Alkaline Phosphatase) Substrate
Western-SuperStar™ Reagent Set (containing Blocker, Wash Buffer, Assay Buffer, Substrate, Enhancer, Blot-Loc™ bags, Development Folders and pair of Tweezers) and an anti-mouse secondary antibody conjugated to alkaline phosphatase
Videos