Learn More
Invitrogen™ TNF alpha Porcine ELISA Kit
Quantifies natural and recombinant swine TNF-α in swine serum, EDTA plasma, buffered solution, or cell culture medium
£478.00 - £616.00
Specifications
Accession Number | P23563 |
---|---|
Assay Range | 23.4-1500 pg/mL |
Assay Sensitivity | <3 pg/mL |
Conjugate | Biotin |
Product Type | ELISA Kit |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10435133
|
Invitrogen™
KSC3011 |
96 Tests |
£478.00
Each |
Estimated Shipment: 03-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10069402
|
Invitrogen™
KSC3012 |
2 x 96 Tests |
£616.00
Each |
Estimated Shipment: 05-08-2024
Log in to see stock. |
|||||
Description
The Porcine Tumor Necrosis Factor alpha (Por TNFα) ELISA quantitates Por TNFα in porcine serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Por TNFα1. Principle of the method The Pociner TNFαsolid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
TNF-α is produced by macrophages and has many immune effects.
Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation & Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet Ice
Specifications
P23563 | |
<3 pg/mL | |
ELISA Kit | |
Porcine | |
Colorimetric Microplate Reader | |
TNFa | |
7.3% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
TNF-α | |
Plasma, 100 μL; Serum, 100 μL; Supernatant, 100 μL | |
Porcine | |
5 hr. |
23.4-1500 pg/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric Detection | |
397086 | |
4 hr. | |
6% | |
HRP | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
For Research Use Only. Not for use in diagnostic procedures.