Learn More
Invitrogen™ Novex TNF- Mouse ELISA Kit
Designed to detect and quantify the level of mouse TNF-α in serum, plasma, and supernatant using 96-well plates and a microplate reader
Brand: Invitrogen™ KMC3011C
Code : 50
Additional Details : Weight : 1.56000kg
Specifications
Colorimetric | |
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers. | |
480 assays | |
Plasma, Serum, Supernatants | |
2.5 hrs |
21926 | |
<3pg/mL | |
RUO - research use only | |
50μL | |
Tumor necrosis factor |
For Research Use Only. Not for use in diagnostic procedures.