missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex TNF- Monkey ELISA Kit
Quantifies natural and recombinant monkey TNF-α in serum, plasma, buffered solution, or cell culture medium
£456.96 - £516.00
Specifications
Sample Type | Plasma, Serum, Supernatant |
---|---|
For Use With (Application) | Colorimetric |
Gene ID (Entrez) | 7124 |
Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
Precision | <2pg/mL |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10384633
|
Invitrogen™
KPC3012 |
192 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10252583
|
Invitrogen™
KPC3011 |
96 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
TNF-α is produced by macrophages and has many immune effects.
Apoptosis, Apoptosis Related Factors, Cell Analysis, Cell Signaling, Cell Viability, Proliferation and Function, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
Plasma, Serum, Supernatant | |
7124 | |
<2pg/mL | |
Tumor necrosis factor |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
For research use only (RUO) | |
4 hr. |
For Research Use Only. Not for use in diagnostic procedures.