Learn More
Invitrogen™ Resistin Human ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KHP0051
Code : 50
Additional Details : Weight : 0.47000kg
Includes: Pre-coated plate, standard, control, detector antibody, anti-rabbit HRP, diluents, wash buffer, chromogen, and stop solution.
Description
The Human Resistin ELISA quantitates Hu Resistin in human serum, plasma, buffered solution, or cell culture medium. The assay will exclusively recognize both natural and recombinant Hu Resistin. Principle of the method The Human Resistin solid-phase sandwich ELISA (enzyme-linked immunosorbent assay) is designed to measure the amount of the target bound between a matched antibody pair. A target-specific antibody has been pre-coated in the wells of the supplied microplate. Samples, standards, or controls are then added into these wells and bind to the immobilized (capture) antibody. The sandwich is formed by the addition of the second (detector) antibody, a substrate solution is added that reacts with the enzyme-antibody-target complex to produce measurable signal. The intensity of this signal is directly proportional to the concentration of target present in the original specimen. Rigorous validation Each manufactured lot of this ELISA kit is quality tested for criteria such as sensitivity, specificity, precision, and lot-to-lot consistency. See manual for more information on validation.
Adipose tissue is an energy reserve in animals and is strictly regulated. Adipose cells produce and secrete numerous physiologically important proteins, such as lipoprotein lipase (LPL), leptin, adipocyte complement related protein of 30 kDa (Acrp30), perilipin, and resistin. Resistin, also known as Adipocyte Secreted Factor (ADSF) and FIZZ3, is a cytokine that is specifically secreted by adipocytes. While the function of resistin is not well understood, it appears to have key paracrine roles and has a suggested role in type 2 diabetes. Studies show that in obese, diabetic mice the levels of serum resistin are increased, which has an antagonistic effect on insulin. Additional studies have shown that resistin causes insulin resistance and glucose intolerance in mice. Resistin gene expression requires C/EBP alpha, which is necessary and sufficient for expression. Studies show that the coactivators CBP and p300, which have been shown to display histone acetyltransferase (HAT) activity, abundantly acetylate the histones at the resistin promoter suggesting their role in activating chromatin for resistin transcription. Numerous PPAR gamma and RXR ligands have been shown to downregulate resistin expression in adipocytes.Order Info
Shipping Condition: Room Temperature
Specifications
Q9HD89 | |
100 pg/mL | |
ELISA Kit | |
Human | |
Colorimetric Microplate Reader | |
ADSF,FIZZ31,RSTN,XCP1,RETN | |
5.6% | |
Pre-coated 96 well plate, Standard, Standard Dilution Buffer, Biotinylated Detection Antibody, Streptavidin-HRP, HRP Diluent, Wash Buffer, Chromogen, Stop Solution, Adhesive Plate Covers | |
Resistin | |
RUO | |
2°C to 8°C | |
1 hr. 20 min. |
0.125-8 ng/mL | |
Biotin | |
Plasma, Serum, Supernatant | |
Colorimetric Detection | |
56729 | |
3.3 hr. | |
3.7% | |
HRP | |
96 Tests | |
Plasma, 25 μL; Serum, 25 μL; Supernatant, 100 μL | |
Human | |
3 hr. 10 min. |
For Research Use Only. Not for use in diagnostic procedures.