Learn More
Invitrogen™ Novex™ MIP-2 Rat ELISA Kit
Quantifies natural and recombinant rat MIP-2 in serum, buffered solution, or cell culture medium
Brand:  Invitrogen™ KRC1022
Code : 50
Additional Details : Weight : 0.68250kg
 View more versions of this product
Product Code. 10486673
  • £528.34 / Each
Estimated Shipment
08-02-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
NM_053647.1, NP_446099.1
114105
3.5 hr.
MIP-2
<1pg/mL
50μL
Colorimetric Detection
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Microplate Reader
192 assays
Serum, Supernatants
chemokine (C-X-C motif) ligand 2
Videos