Learn More
Invitrogen™ Novex™ MCP-1 Rat ELISA Kit
Quantifies natural and recombinant rat Monocyte Chemoattractant Protein-1 (MCP-1) in serum, plasma, buffered solution, or cell culture medium
Brand:  Invitrogen™ KRC1012
Code : 50
Additional Details : Weight : 0.65000kg
 View more versions of this product
Product Code. 10089362
  • £458.71 / Each
Estimated Shipment
12-03-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
NM_031530.1, NP_113718.1
24770
2.5 hr.
MCP-1
<8pg/mL
50μL
Colorimetric Detection
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Microplate Reader
192 assays
Plasma, Serum, Supernatants
chemokine (C-C motif) ligand 2
Videos