Learn More
Invitrogen™ Novex™ Leukotriene B4 Human ELISA Kit
Competitive immunoassay to quantify LTB4 in biological samples
Brand: Invitrogen™ KHL1741
Code : 50
Additional Details : Weight : 0.15000kg
Includes: Pre-coated plate, standard, antibody, AP tracer, wash buffer, tris buffer, DEA buffer, pNPP tablets, and plate cover.
Description
LTB4 is a potent inflammatory mediator that works by binding BLT1, on leukocytes.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Room Temperature
Specifications
NM_001143919.2, NM_181657.3, NP_001137391.1, NP_858043.1 | |
Colorimetric Detection | |
1241 | |
Antibody, AP tracer, DEA buffer, Plate covers, pNPP tablets, Pre-coated plate, Standard, Tris buffer, Wash buffer | |
96 Assays |
Plasma, Serum | |
Microplate Reader | |
3 hr. | |
Leukotriene B4 | |
leukotriene B4 receptor |
For Research Use Only. Not for use in diagnostic procedures.