Invitrogen™ Novex IL-6 Rat ELISA Kit

Cytokine with actions in immune response

Overview
Brand: Invitrogen™

Manufacturer Part Number: KRC0062

UNSPSC: 12352203

Code: NEW

Additional Details:
Additional Details: Weight: 0.65000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15133637

Quantity Price
1 £ 448.0 / Each
EU Stock 2
Add to basket


Description and Specification

Specification

Regulatory Status RUO - research use only
Sample Size 50.0μL
Target Interleukin 6
Precision <5pg/mL
Sample Type Serum, Supernatants
Quantity 192 assays
For Use With (Application) Colorimetric
Includes Pre-coated plate, standard, detector antibody, anti-IgG-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Gene ID (Entrez) 24498
Test Time 4.5 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.