missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Novex IL-4 Swine ELISA Kit
Key cytokine in the immune network
£358.50 - £458.71
Specifications
Sample Type | Serum, Supernatant |
---|---|
For Use With (Application) | Colorimetric |
Gene ID (Entrez) | 397225 |
Kit Contents | Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer |
Precision | 2pg/mL |
Includes: Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10671314
|
Invitrogen™
KSC0041 |
96 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
10454383
|
Invitrogen™
KSC0042 |
192 Assays |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
The IL-4 Swine ELISA Kit quantifies natural and recombinant swine IL-4 in serum, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Order Info
Shipping Condition: Wet ice
Specifications
Serum, Supernatant | |
397225 | |
2pg/mL | |
Interleukin 4 |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
For research use only (RUO) | |
3 hr. |
For Research Use Only. Not for use in diagnostic procedures.