Learn More
Invitrogen™ Novex IL-4 Swine ELISA Kit
Key cytokine in the immune network
Brand: Invitrogen™ KSC0042
Code : 50
Additional Details : Weight : 0.68250kg
Description
The IL-4 Swine ELISA Kit quantifies natural and recombinant swine IL-4 in serum, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Serum, Supernatant | |
397225 | |
2pg/mL | |
For research use only (RUO) | |
3 hr. |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
192 Assays | |
Interleukin 4 |
For Research Use Only. Not for use in diagnostic procedures.