Learn More
Invitrogen™ Novex IL-4 Human Ultrasensitive ELISA Kit
Brand:  Invitrogen™ KHC0044
Code : 50
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 10079122
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Product Support team on 01509 555888 for assistance
View alternative products »
Specifications
Specifications
Colorimetric
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
96 assays
Plasma, Serum, Supernatants
3.5 hrs
3565
<0.27pg/mL
RUO - research use only
100μL
Interleukin 4
Videos