Learn More
Invitrogen™ Novex IL-2 Rat ELISA Kit
Promotes T-cell growth
Brand: Invitrogen™ KRC0021
Code : 50
Additional Details : Weight : 0.70000kg
Description
The IL-2 Rat ELISA Kit quantifies natural and recombinant rat IL-2 in serum, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Serum, Supernatant | |
116562 | |
<5pg/mL | |
For research use only (RUO) | |
3 hr. |
Colorimetric | |
Chromogen, Detector antibody, Diluents, Plate covers, Pre-coated plate, Standard, Stop solution, Streptavidin-HRP, Wash buffer | |
96 Assays | |
Interleukin 2 |