Learn More
Invitrogen™ Novex IL-2 Mouse ELISA Kit
Designed to detect and quantify the level of mouse IL-2 in serum, plasma, and supernatant using 96-well plates and a microplate reader
Brand:  Invitrogen™ KMC0022
Code : 50
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10252963
  • £462.81 / Each
Estimated Shipment
11-06-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Specifications
Specifications
Colorimetric
Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.
192 assays
Plasma, Serum, Supernatants
3 hrs
16183
<8pg/mL
RUO - research use only
50μL
Interleukin 2