Invitrogen™ Novex IL-2 Monkey ELISA Kit

Promotes T-cell growth

Overview
Brand: Invitrogen™

Manufacturer Part Number: KPC0022

UNSPSC: 12352203

Code: 50

Additional Details:
Additional Details: Weight: 0.65000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10533195

Quantity Price
1 £ 438.0 / Each
Estimated Shipment date
from Supplier 12-04-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Regulatory Status RUO - research use only
Sample Type Serum, Supernatants
Sample Size 50μL
Target Interleukin 2
Precision <2pg/mL
For Use With (Application) Colorimetric
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Quantity 192 assays
Gene ID (Entrez) 3558
Test Time 4 hrs

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.