Learn More
Invitrogen™ IL-1 beta Mouse ELISA Kit
Sandwich ELISA Kit
Brand: Invitrogen™ KMC0012
Code : 50
Additional Details : Weight : 0.65000kg
Description
The IL-1β Mouse ELISA Kit quantifies natural and recombinant mouse IL-1β in serum, plasma, buffered solution, or cell culture medium.
Cell Analysis, Immunoassays, Ready-To-Use Immunoassay
Specifications
Plasma, Serum, Supernatant | |
16176 | |
<7pg/mL | |
For research use only (RUO) | |
2.75 hr. |
Colorimetric | |
Chromogen, Controls, Detector antibody, Detector antibody, Incubation buffer, Incubation buffer, Pre-coated plate, Pre-coated plate, Standard, Streptavidin-HRP, Streptavidin-HRP | |
192 Assays | |
Interleukin 1β |