Learn More
Invitrogen™ Novex IL-1B Mouse ELISA Kit
Important in immune response
Brand:  Invitrogen™ KMC0011
Code : 50
Additional Details : Weight : 0.47000kg
 View more versions of this product
Product Code. 10141113
  • £534.15 / Each
EU Stock 5
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Colorimetric
Pre-coated plate, standard, controls, detector antibody, Streptavidin-HRP, diluents, wash buffer, incubation buffer, chromogen, stop solution, and plate covers.
96 assays
Plasma, Serum, Supernatants
2.75 hrs
16176
<7pg/mL
RUO - research use only
50μL
Interleukin 1β
Videos