Invitrogen™ Novex™ IL-15 Human ELISA Kit

Quantifies natural and recombinant human IL-15 in serum, buffered solution, or cell culture medium

Overview
Brand: Invitrogen™

Manufacturer Part Number: KHC0152

UNSPSC: 12352200

Code: 50

Additional Details:
Additional Details: Weight: 0.65000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10745773

Quantity Price
1 £ 438.0 / 192 tests
Estimated Shipment date
from Supplier 13-03-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Sensitivity <10pg/mL
Incubation Time 2 hr.
Marker Gene IL-15
Accession Number NM_000585.4, NM_172174.2, NM_172175.2, NP_000576.1, NP_751915.1
For Use With (Equipment) Microplate Reader
Sample Size 50μL
Sample Type Serum, Supernatants
Target interleukin 15
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers
For Use With (Application) Colorimetric Detection
Quantity 192 assays
Gene ID (Entrez) 3600

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.