Invitrogen™ Novex IL-13 Human Ultrasensitive ELISA Kit

A cytokine produced primarily by Th2 cells

Overview
Brand: Invitrogen™

Manufacturer Part Number: KHC0133

UNSPSC: 12352203

Code: 50

Additional Details:
Additional Details: Weight: 0.50000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 10079272

Quantity Price
1 £ 520.0 / 192 tests
Estimated Shipment date
from Supplier 11-05-2017
Add to basket

For Research Use Only. All usage must comply with product instructions.
 

Description and Specification

Specification

Regulatory Status RUO - research use only
Precision <0.6pg/mL
Sample Type Plasma, Serum, Supernatants
Target Interleukin 13
Sample Size 50μL
Quantity 192 assays
For Use With (Application) Colorimetric
Includes Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
Test Time 4 hrs
Gene ID (Entrez) 3596

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.