Learn More
Invitrogen™ Novex IL-12 Human ELISA Kit
Quantifies natural and recombinant human IL-12 p35/p40 heterodimer (or IL-12p70)
Brand:  Invitrogen™ KHC0122
Code : 50
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 10766713
  • £458.71 / 192 tests
EU Stock 1
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Specifications
Colorimetric
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
192 assays
Plasma, Serum, Supernatants
3 hrs
3592
<2pg/mL
RUO - research use only
50μL
Interleukin 12A
Videos