Learn More
Invitrogen™ Novex IL-10 Rat ELISA Kit
Lymphokine involved in immune response
Brand:  Invitrogen™ KRC0101
Code : 50
Additional Details : Weight : 0.70000kg
 View more versions of this product
Product Code. 10304453
  • £361.58 / Each
Estimated Shipment
14-06-2018
Add to basket
For Research Use Only. All usage must comply with product instructions.
Specifications
Specifications
Colorimetric
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
96 assays
Serum, Supernatants
3 hrs
25325
<5pg/mL
RUO - research use only
50μL
Interleukin 10