Learn More
Invitrogen™ Novex IL-10 Mouse ELISA Kit
A lymphokine involved in immune response
Brand:  Invitrogen™ KMC0102
UNSPSC : 12352203
Code : 50
Additional Details : Weight : 0.65000kg
 View more versions of this product
Product Code. 10652644
  • £452.00 / Each
Estimated Shipment
11-08-2017
Add to basket
For Research Use Only. All usage must comply with product instructions.
Disclaimers:
For Research Use Only. Not for use in diagnostic procedures.
Specifications Description & Specifications
Specifications
Colorimetric
Pre-coated plate, standard, detector antibody, Streptavidin-HRP, diluents, wash buffer, chromogen, stop solution, and plate covers.
192 assays
Serum, Supernatants
3 hrs
16153
<13pg/mL
RUO - research use only
100μL
Interleukin 10